missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green - Methyl Red, Alcoholic, for Alkalinity, Certified, LabChem™

Supplier:  LabChem LC118807

 View more versions of this product

Catalog No. LC118807


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bromocresol Green - Methyl Red
Alcoholic, for Alkalinity
90.36,5,4.5,0.1,0.04
C21H14Br4O5S·Na
Soluble in water
720.06
White Poly
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Green
13°C
-115°C
78°C
67-63-0,67-56-1,62625-32-5,845-10-3
UN1170
Passes Test
Certified
790kg/m3
790kg/m3
125 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions