missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green - Methyl Red, Alcoholic, for Alkalinity, Certified, LabChem™
Supplier: LabChem LC118801
Specifications
| Bromocresol Green - Methyl Red | |
| Alcoholic, for Alkalinity | |
| 90.36,5,4.5,0.1,0.04 | |
| C21H14Br4O5S·Na | |
| Soluble in water | |
| 720.06 | |
| Amber Poly | |
| Fume; Carbon monoxide; Carbon dioxide; May release flammable gases | |
| Green | |
| 13°C |
| -115°C | |
| 78°C | |
| 67-63-0,67-56-1,62625-32-5,845-10-3 | |
| UN1170 | |
| Passes Test | |
| Certified | |
| 790kg/m3 | |
| 790kg/m3 | |
| 500 mL | |
| Liquid |