missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Indicator, 0.1% (w/v), Ricca Chemical

Catalog No. R12050001A
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R12050001A 1 L Natural Poly Bottle
12054 120 mL Natural Poly Bottle
120516 500 mL Natural Poly Bottle
12051 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R12050001A Supplier Ricca Chemical Company Supplier No. R12050001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-32-5

Specifications

Chemical Name or Material Bromocresol Green Indicator
CAS 62625-32-5, 7732-18-5
CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Concentration 0.1% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Dark Blue
pH >5.4
Quantity 1 L
Physical Form Liquid
Percent Purity 0.1% W/V
Grade Indicator
Show More Show Less