missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, 0.1% (w/v) Aqueous Indicator Solution, Spectrum™ Chemical

Catalog No. 18609226
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18609226 125 mL
18609227 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18609226 Supplier Spectrum Chemical Mfg Cor Supplier No. B163125ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Specifications

Chemical Name or Material Bromocresol Green
CAS 62625-32-5, 7732-18-5
Concentration 0.10%
Grade Chemical Solution
Packaging Poly Bottle
Color Blue/Green
Quantity 125 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.