missing translation for 'onlineSavingsMsg'
Learn More

Bromocholine Bromide 99.0+%, TCI America™

Catalog No. B057725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B057725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B057725G Supplier TCI America Supplier No. B057725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2758-06-7
Molecular Formula C5H13Br2N
Molecular Weight (g/mol) 246.97
MDL Number MFCD00011865
InChI Key OINMNSFDYTYXEQ-UHFFFAOYSA-M
Synonym 2-bromoethyl trimethylammonium bromide, 2-bromo-n,n,n-trimethylethanaminium bromide, 2-bromoethyltrimethylammonium bromide, bromcholin, hypercyl, kathesin, btab, btab the plant regulator, bcb the plant regulator, unii-kh1i7ru18l
PubChem CID 17689
IUPAC Name 2-bromoethyl(trimethyl)azanium;bromide
SMILES C[N+](C)(C)CCBr.[Br-]

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 246.97
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bromocholine Bromide
RTECSNumber BO9481000
TSCA Yes