missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl Chloride 85.0+%, TCI America™

Catalog No. p-7118732
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B090025G 25 g
B0900500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B090025G Supplier TCI America Supplier No. B090025G

Chemical Identifiers

CAS 22118-09-8
Molecular Formula C2H2BrClO
Molecular Weight (g/mol) 157.391
MDL Number MFCD00000724
InChI Key SYZRZLUNWVNNNV-UHFFFAOYSA-N
Synonym bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride
PubChem CID 89602
IUPAC Name 2-bromoacetyl chloride
SMILES C(C(=O)Cl)Br

Specifications

Color Yellow
Boiling Point 128°C
Quantity 25 g
UN Number 3265
Formula Weight 157.39
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoacetyl Chloride
TSCA No
Recommended Storage Refrigerator