Learn More
Bromoacetyl chloride, 95%
CAS: 22118-09-8 | C2H2BrClO | 157.39 g/mol
$137.72 - $137.72
Chemical Identifiers
| CAS | 22118-09-8 |
|---|---|
| Molecular Formula | C2H2BrClO |
| Molecular Weight (g/mol) | 157.39 |
| MDL Number | MFCD00000724 |
| InChI Key | SYZRZLUNWVNNNV-UHFFFAOYSA-N |
| Synonym | bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride |
| PubChem CID | 89602 |
| IUPAC Name | 2-bromoacetyl chloride |
| SMILES | C(C(=O)Cl)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC197990250
|
Thermo Scientific Chemicals
197990250 |
25 g | Glass bottle |
Each for $137.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 22118-09-8 | |
| 157.39 | |
| SYZRZLUNWVNNNV-UHFFFAOYSA-N | |
| 89602 | |
| C(C(=O)Cl)Br |
| C2H2BrClO | |
| MFCD00000724 | |
| bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride | |
| 2-bromoacetyl chloride |
Specifications
| 22118-09-8 | |
| 1.9000g/mL | |
| Authentic | |
| Glass bottle | |
| 1.496 | |
| MFCD00000724 | |
| 02,215 | |
| bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride | |
| SYZRZLUNWVNNNV-UHFFFAOYSA-N | |
| 2-bromoacetyl chloride | |
| 89602 | |
| 95% | |
| Bromoacetyl chloride |
| Yellow to Brown | |
| 127.0°C to 128.0°C | |
| 94% min. (GC) | |
| C2H2BrClO | |
| BrCH2COCl | |
| 25 g | |
| 1.9 | |
| Solubility in water: reacts | |
| C(C(=O)Cl)Br | |
| 157.39 | |
| 157.39 | |
| Liquid |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 244-790-7
Recommended Storage : 4°C
RUO – Research Use Only