missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl bromide, 98%

Catalog No. p-72126
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106601000 100 mL Glass bottle
AC106605000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106601000 Supplier Thermo Scientific Chemicals Supplier No. 106601000
Only null left

CAS: 598-21-0 | C2H2Br2O | 201.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-21-0
Molecular Formula C2H2Br2O
Molecular Weight (g/mol) 201.85
MDL Number MFCD00000115
InChI Key LSTRKXWIZZZYAS-UHFFFAOYSA-N
Synonym bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
PubChem CID 61147
IUPAC Name 2-bromoacetyl bromide
SMILES C(C(=O)Br)Br

Specifications

Color White
Density 2.3100g/mL
Boiling Point 147°C to 150°C
Flash Point >105°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.547
Linear Formula BrCH2COBr
Quantity 100 mL
Beilstein 02, 215
Specific Gravity 2.31
Solubility Information Solubility in water: reacts. Other solubilities: solulbe in acetone,carbontetrachloride
Formula Weight 201.85
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bromoacetyl bromide, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 209-923-5
TSCA TSCA
Recommended Storage May discolor to brown on storage

RUO – Research Use Only