missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic anhydride, 95%

Catalog No. AC426700250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426700250 25 g Glass bottle
AC426700050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426700250 Supplier Thermo Scientific Chemicals Supplier No. 426700250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13094-51-4 | C4H4Br2O3 | 259.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13094-51-4
Molecular Formula C4H4Br2O3
Molecular Weight (g/mol) 259.87
MDL Number MFCD00045041
InChI Key FUKOTTQGWQVMQB-UHFFFAOYSA-N
Synonym bromoacetic anhydride, 2-bromoacetic anhydride, acetic acid, 2-bromo-,1,1'-anhydride, 2-bromoacetyl 2-bromoacetate, bromoaceticanhydride, acetic acid, bromo-, anhydride
PubChem CID 4259817
IUPAC Name (2-bromoacetyl) 2-bromoacetate
SMILES C(C(=O)OC(=O)CBr)Br

Specifications

Melting Point 40°C to 44°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Br) (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2C(=O)OC(=O)CH2Br
Quantity 25 g
Formula Weight 259.87
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Bromoacetic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only