missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic Acid 98.0+%, TCI America™

Catalog No. B0531500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0531500G 500 g
B053125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0531500G Supplier TCI America Supplier No. B0531500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-08-3
Molecular Formula C2H3BrO2
Molecular Weight (g/mol) 138.948
MDL Number MFCD00002678
InChI Key KDPAWGWELVVRCH-UHFFFAOYSA-N
Synonym bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
PubChem CID 6227
IUPAC Name 2-bromoacetic acid
SMILES C(C(=O)O)Br

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 208°C
Quantity 500 g
UN Number 3425
Formula Weight 138.95
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bromoacetic Acid
EINECSNumber (2)-2634
RTECSNumber AF5950000
TSCA Yes