missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid, 98+%

Catalog No. AAA1440330
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1440330 250 g
AAA144030B 1000 g
AAA144030I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1440330 Supplier Thermo Scientific Chemicals Supplier No. A1440330

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-08-3 | C2H3BrO2 | 138.948 g/mol

Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water, alcohol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to light and air. Handle and store under inert gas. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 79-08-3
Molecular Formula C2H3BrO2
Molecular Weight (g/mol) 138.948
MDL Number MFCD00002678
InChI Key KDPAWGWELVVRCH-UHFFFAOYSA-N
Synonym bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
PubChem CID 6227
IUPAC Name 2-bromoacetic acid
SMILES C(C(=O)O)Br

Specifications

Melting Point 44°C to 49°C
Density 1.93
Boiling Point 207°C to 209°C
Flash Point >110°C (230°F)
Odor Pungent
Linear Formula BrCH2CO2H
Quantity 250 g
UN Number UN3425
Beilstein 506167
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,1400
Solubility Information Soluble in water, alcohol
Formula Weight 138.95
Percent Purity ≥98%
Chemical Name or Material Bromoacetic acid
Show More Show Less
Hazard Category H301+H311+H331-H314-H317-H335
Hazard Statement GHS H Statement
H301-H311-H331-H314-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BROMOACETIC ACID, SOLID
EINECSNumber 201-175-8
RTECSNumber AF5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only