missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Bromoacetic Acid

Catalog No. 17000500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17000500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000500G Supplier Sigma Aldrich Supplier No. 17000500G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bromoacetic Acid

Specifications

CAS 79-08-3
Melting Point 46°C to 49°C; 47°C to 49°C
Boiling Point 208°C
Molecular Formula C2H3BrO2
Linear Formula BrCH2COOH
MDL Number MFCD00002678
Quantity 500 g
Synonym alpha-Bromoacetic acid; Monobromoacetic acid
Molecular Weight (g/mol) 138.95 g/mol