missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde Dimethyl Acetal (stabilized with K2CO3) 97.0+%, TCI America™

Catalog No. B1097500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1097500G 500 g
B109725G 25 g
B1097100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1097500G Supplier TCI America Supplier No. B1097500G

Chemical Identifiers

CAS 7252-83-7
Molecular Formula C4H9BrO2
Molecular Weight (g/mol) 169.018
MDL Number MFCD00000213
InChI Key FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Synonym bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
PubChem CID 81672
IUPAC Name 2-bromo-1,1-dimethoxyethane
SMILES COC(CBr)OC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 500 g
UN Number 1993
Formula Weight 169.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoacetaldehyde Dimethyl Acetal (stabilized with K2CO3)