missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde dimethyl acetal, 97%, stabilized

Catalog No. AC221870250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221870250 25 mL Glass bottle
AC221871000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221870250 Supplier Thermo Scientific Chemicals Supplier No. 221870250

CAS: 7252-83-7 | C4H9BrO2 | 169.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7252-83-7
Molecular Formula C4H9BrO2
Molecular Weight (g/mol) 169.02
MDL Number MFCD00000213
InChI Key FUSFWUFSEJXMRQ-UHFFFAOYSA-N
Synonym bromoacetaldehyde dimethyl acetal, ethane, 2-bromo-1,1-dimethoxy, 2,2-dimethoxyethyl bromide, dimethylbromoacetal, 1-bromo-2,2-dimethoxyethane, 2,2-dimethoxybromoethane, 2-bromoacetaldehyde dimethyl acetal, bromoacetoaldehyde dimethyl acetal, bromoacetaldehyde dimethylacetal, acetaldehyde, bromo-, dimethyl acetal
PubChem CID 81672
IUPAC Name 2-bromo-1,1-dimethoxyethane
SMILES COC(CBr)OC

Specifications

Density 1.4300g/mL
Boiling Point 148.0°C to 150.0°C
Flash Point 51°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.4470
Linear Formula BrCH2CH(OCH3)2
Quantity 25 mL
Beilstein 01, 624
Specific Gravity 1.43
Solubility Information Solubility in water: insoluble
Formula Weight 169.02
Percent Purity 97%
Chemical Name or Material Bromoacetaldehyde dimethyl acetal, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Fatal if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
EINECSNumber 230-669-6
TSCA TSCA

RUO – Research Use Only