missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoacetaldehyde Diethyl Acetal 95.0+%, TCI America™
$78.93 - $594.89
Chemical Identifiers
| CAS | 2032-35-1 |
|---|---|
| Molecular Formula | C6H13BrO2 |
| Molecular Weight (g/mol) | 197.07 |
| MDL Number | MFCD00000214 |
| InChI Key | LILXDMFJXYAKMK-UHFFFAOYSA-N |
| Synonym | bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal |
| PubChem CID | 74852 |
| IUPAC Name | 2-bromo-1,1-diethoxyethane |
| SMILES | CCOC(CBr)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B095725ML
|
TCI America
B095725ML |
25 mL |
Each for $78.93
|
|
|||||
|
B0957100ML
|
TCI America
B0957100ML |
100 mL |
Each for $198.30
|
|
|||||
|
B0957500ML
|
TCI America
B0957500ML |
500 mL |
Each for $594.89
|
|
|||||
Chemical Identifiers
| 2032-35-1 | |
| 197.07 | |
| LILXDMFJXYAKMK-UHFFFAOYSA-N | |
| 74852 | |
| CCOC(CBr)OCC |
| C6H13BrO2 | |
| MFCD00000214 | |
| bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal | |
| 2-bromo-1,1-diethoxyethane |
Specifications
| 2032-35-1 | |
| 67°C | |
| MFCD00000214 | |
| 1992 | |
| LILXDMFJXYAKMK-UHFFFAOYSA-N | |
| 2-bromo-1,1-diethoxyethane | |
| 74852 | |
| ≥95.0% (GC) | |
| Bromoacetaldehyde Diethyl Acetal |
| Yellow | |
| C6H13BrO2 | |
| 25 mL | |
| bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal | |
| CCOC(CBr)OCC | |
| 197.07 | |
| 197.07 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2467
TSCA : Yes