missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde Diethyl Acetal 95.0+%, TCI America™

Catalog No. B095725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B095725ML 25 mL
B0957100ML 100 mL
B0957500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B095725ML Supplier TCI America Supplier No. B095725ML

Chemical Identifiers

CAS 2032-35-1
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.07
MDL Number MFCD00000214
InChI Key LILXDMFJXYAKMK-UHFFFAOYSA-N
Synonym bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
PubChem CID 74852
IUPAC Name 2-bromo-1,1-diethoxyethane
SMILES CCOC(CBr)OCC

Specifications

Color Yellow
Boiling Point 67°C
Quantity 25 mL
UN Number 1992
Formula Weight 197.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoacetaldehyde Diethyl Acetal
EINECSNumber (2)-2467
TSCA Yes