missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde diethyl acetal, 97%

Catalog No. AAA1327622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1327622 100 g
AAA1327636 500 g
AAA132760E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1327622 Supplier Thermo Scientific Chemicals Supplier No. A1327622
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 2032-35-1 | C6H13BrO2 | 197.072 g/mol

Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ?-heterosubstituted acetaldehyde acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ɑ-heterosubstituted acetaldehyde acetals.

Solubility
It is immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Protect from frost, heat and sunlight. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Acids. Moisture Sensitive.

Chemical Identifiers

CAS 2032-35-1
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.07
MDL Number MFCD00000214
InChI Key LILXDMFJXYAKMK-UHFFFAOYSA-N
Synonym bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
PubChem CID 74852
IUPAC Name 2-bromo-1,1-diethoxyethane
SMILES CCOC(CBr)OCC

Specifications

Density 1.27
Boiling Point 66°C to 67°C (18 mmHg)
Flash Point 61°C (142°F)
Refractive Index 1.439
Linear Formula BrCH2CH(OCH2CH3)2
Quantity 100 g
UN Number UN2810
Beilstein 635754
Sensitivity Moisture sensitive
Solubility Information It is immiscible with water.
Formula Weight 197.08
Percent Purity 97%
Chemical Name or Material Bromoacetaldehyde diethyl acetal
Show More Show Less
Hazard Category H227-H301-H315-H319-H330
Hazard Statement GHS H Statement
H301-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 217-989-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only