missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde diethyl acetal, 97%

CAS: 2032-35-1 | C6H13BrO2 | 197.07 g/mol

$90.75 - $359.72

Chemical Identifiers

CAS 2032-35-1
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.07
MDL Number MFCD00000214
InChI Key LILXDMFJXYAKMK-UHFFFAOYSA-N
Synonym bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
PubChem CID 74852
IUPAC Name 2-bromo-1,1-diethoxyethane
SMILES CCOC(CBr)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC346521000
SDS View Documents
Thermo Scientific Chemicals
346521000
100 mL Glass bottle
Each for $90.75
Only null left
 
AC346525000
SDS View Documents
Thermo Scientific Chemicals
346525000
500 mL Glass bottle
Each for $359.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2032-35-1
197.07
LILXDMFJXYAKMK-UHFFFAOYSA-N
74852
CCOC(CBr)OCC
C6H13BrO2
MFCD00000214
bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
2-bromo-1,1-diethoxyethane

Specifications

2032-35-1
66.0°C to 67.0°C (18.0 mmHg)
96% min. (GC)
C6H13BrO2
BrCH2CH(OC2H5)2
100 mL
1.28
Solubility in water: immiscible
CCOC(CBr)OCC
197.07
197.07
Bromoacetaldehyde diethyl acetal, 97%
1.2800g/mL
61°C
Glass bottle
1.4380 to 1.4400
MFCD00000214
01,625
bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
LILXDMFJXYAKMK-UHFFFAOYSA-N
2-bromo-1,1-diethoxyethane
74852
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Fatal if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 217-989-1

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only