missing translation for 'onlineSavingsMsg'
Learn More

Bromo(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7051334
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
AA4100203 1 g
1 options

Catalog No. AA4100203

Supplier: Thermo Scientific Chemicals 04100203

Only null left

Bromo(triphenylphosphine)gold(I) is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14243-65-3
Molecular Formula C18H15AuBrP
Molecular Weight (g/mol) 539.16
MDL Number MFCD00049267
InChI Key UIYUJCNGYSSFGP-UHFFFAOYSA-M
Synonym bromo triphenylphosphine gold i, bromoaurio triphenyl-??-phosphanyl
PubChem CID 12464140
IUPAC Name bromogold;triphenylphosphane
SMILES Br[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 539.17
Percent Purity 99.99%
Grade Premion
Assay (metals basis)
Chemical Name or Material Bromo(triphenylphosphine)gold(I)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-118-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only