missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromo[(2,6-pyridinediyl)bis(3-methyl-1-imidazolyl-2-ylidene)]nickel Bromide 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 457.78 |
| Percent Purity | ≥98.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bromo[(2,6-pyridinediyl)bis(3-methyl-1-imidazolyl-2-ylidene)]nickel Bromide |
Safety and Handling
| TSCA | No |