missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, 99.99+% (metals basis)

Catalog No. p-4407290
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA87614AA 5 mL
AA87614AD 50 mL
AA87614AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA87614AA Supplier Thermo Scientific Chemicals Supplier No. 087614AA
Only null left

CAS: 7726-95-6 | Br | 79.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]

Specifications

Color Red to Brown
Physical Form Liquid
Quantity 5 mL
Assay Percent Range 99.998% (metals basis)
IUPAC Name bromide
Percent Purity 99.99+%
Odor Unpleasant
Vapor Pressure 173mm Hg at 20°C
Chemical Name or Material Bromine liquid
Hazard Category H314-H330
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: I
EINECSNumber 231-778-1
RTECSNumber EF9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only