missing translation for 'onlineSavingsMsg'
Learn More

Bromine, 99.6%, for analysis

CAS: 7726-95-6 | Br | 79.91 g/mol

$1677.62 - $1677.62

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC196660010
SDS View Documents
Thermo Scientific Chemicals
196660010
1 L
Each for $1,677.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent for analysis
Specifications

Chemical Identifiers

7726-95-6
79.91
CPELXLSAUQHCOX-UHFFFAOYSA-M
24408
[Br-]
Br
MFCD00010896
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CHEBI:29224

Specifications

58.7°C
7726-95-6
99.5% min. (Iodometry)
Br2
03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CPELXLSAUQHCOX-UHFFFAOYSA-M
bromide
24408
159.82
Analytical
3.111
Bromine, p.a.
-7.2°C
1 L
Br
MFCD00010896
15, 1401
Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
[Br-]
79.91
CHEBI:29224
99.6%
Plastic Coated Glass Bottle
3.1110g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-778-1

RTECSNumber : EF9100000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only