missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 58.7°C |
| Chemical Name or Material | Bromine |
| Melting Point | -7.2°C |
| Name Note | 1M Solution in trimethyl Phosphate |
| CAS | 7726-95-6, 512-56-1 |
| Quantity | 100 mL |
| Flash Point | 113°C |
| Packaging | AcroSeal™ Glass Bottle |
| Specific Gravity | 1.307 |
| Density | 1.3070g/mL |
| Show More |
RUO – Research Use Only