missing translation for 'onlineSavingsMsg'
Learn More

Bromelain, sourced from stem, >=2500 GDU/g (>=5.0 FIP U/mg)

Catalog No. AC448210250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448210250 25 g
AC448211000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448210250 Supplier Thermo Scientific Chemicals Supplier No. 448210250
Only null left

CAS: 37189-34-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37189-34-7
MDL Number MFCD00081423

Specifications

Ignition Residue 5% max
Packaging Glass bottle
Quantity 25 g
Merck Index 15,1395
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether
Chemical Name or Material Bromelain, 2500 GDU/g
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 253-387-5

RUO – Research Use Only