missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brilliant Yellow, TCI America™
Description
- Appearance: Orange to brown powder to crystal
- Formula Weight: 624.55 g/mol
- Solubility: Soluble in water
- Maximum Absorption Wavelength: 492 nm (0.01mol/L NaOH sol.)
- Lambda max.: 488.0 to 494.0 nm (0.01mol/L NaOH sol.)
- Absorbance (E1%1cm): 810 min.
- NMR: Confirmed to structure
- Storage Temperature: Room temperature (recommended in a cool and dark place, <15°C)
- Applications: Laboratory reagent
Specifications
Specifications
| Color | Yellow to Orange |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 624.55 |
| Grade | Reagent |
| Chemical Name or Material | Brilliant Yellow |
Safety and Handling
| EINECSNumber | (4)-0294 |
| TSCA | Yes |