missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Crocein

CAS: 5413-75-2 | C22H16N4Na2O7S2 | 558.491 g/mol

Supplier:  Thermo Scientific Chemicals A1464218

Quantity 50 g

 View more versions of this product

Catalog No. AAA1464218


Only null left

Description

Description

Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

5413-75-2
558.491
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
88641610
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
C22H16N4Na2O7S2
MFCD00003907
C.I. 27290; Crocein Scarlet MOO
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium

Specifications

Brilliant Crocein
C22H16N4Na2O7S2
C.I. 27290; Crocein Scarlet MOO
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
88641610
50 g
5413-75-2
MFCD00003907
Soluble in water
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
558.491
556.49
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-502-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions