missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brilliant Blue R, TCI America™

Manufacturer:  Tci America C070025G

Catalog No. C070025G



Description

Description

Specifications

Specifications

Brilliant Blue R
C45H44N3NaO7S2
Acid Cyanine 6B, Alizarin Rubinol 5G, Solar Cyanine 6B, Acid Blue 83, Coomassie Brilliant Blue R-250, CBB R-250
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
825.97
825.97
Black-Purple
6104-59-2
MFCD00041762
HAEGGLCXLFWTBE-UHFFFAOYSA-M
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
61365
Crystal-Powder at 20°C
25g

Chemical Identifiers

6104-59-2
825.97
HAEGGLCXLFWTBE-UHFFFAOYSA-M
61365
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
C45H44N3NaO7S2
MFCD00041762
Acid Cyanine 6B, Alizarin Rubinol 5G, Solar Cyanine 6B, Acid Blue 83, Coomassie Brilliant Blue R-250, CBB R-250
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1655

TSCA : Yes

SDS
Documents