Learn More
Brilliant Blue R soln., Ready-to-Use, Thermo Scientific Chemicals
$215.88 - $215.88
Chemical Identifiers
| CAS | 6104-59-2 |
|---|---|
| Molecular Formula | C45H44N3NaO7S2 |
| Molecular Weight (g/mol) | 825.97 |
| MDL Number | MFCD00041762 |
| InChI Key | HAEGGLCXLFWTBE-UHFFFAOYSA-M |
| Synonym | Coomassie Brilliant Blue R; C.I. 42660 |
| PubChem CID | 61365 |
| IUPAC Name | sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate |
| SMILES | [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61384K2
|
Thermo Scientific Chemicals
J61384K2 |
1 L |
Each for $215.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6104-59-2 | |
| 825.97 | |
| HAEGGLCXLFWTBE-UHFFFAOYSA-M | |
| 61365 | |
| [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1 |
| C45H44N3NaO7S2 | |
| MFCD00041762 | |
| Coomassie Brilliant Blue R; C.I. 42660 | |
| sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate |
Specifications
| 6104-59-2 | |
| MFCD00041762 | |
| 5718025 | |
| HAEGGLCXLFWTBE-UHFFFAOYSA-M | |
| sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate | |
| 61365 | |
| Blue | |
| Liquid |
| C45H44N3NaO7S2 | |
| UN1992 | |
| Coomassie Brilliant Blue R; C.I. 42660 | |
| [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1 | |
| 825.97 | |
| 825.97 | |
| 1 L | |
| Brilliant Blue R soln., Ready-to-Use |
Safety and Handling
GHS H Statement
H311-H331-H370-H226
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Flammable liquid and vapor.
GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H500
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 228-060-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only