missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Blue R, pure

CAS: 6104-59-2 | C45H44N3NaO7S2 | 825.97 g/mol

Supplier:  Thermo Scientific Chemicals 191490250

 View more versions of this product

Catalog No. AC191490250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

700 (at lambda min.)
174.0°C to 180.0°C
6104-59-2
C45H44N3NaO7S2
Acid Blue 83, C.I. 42660, Coomassie™ Brilliant Blue R-250
HAEGGLCXLFWTBE-UHFFFAOYSA-M
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
825.97
588 to 596nm (in 50:50 ethanol:water)
Pure
Bordeaux to Brown/Purple
Crystalline Powder
Brilliant Blue R
Pure
1 to 1.2
MFCD00041762
Solubility in water: slightly soluble. Other solubilities: practically insoluble in ethanol
Authentic
sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
61365
825.98
Glass bottle
25 g

Chemical Identifiers

6104-59-2
825.97
HAEGGLCXLFWTBE-UHFFFAOYSA-M
61365
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
C45H44N3NaO7S2
MFCD00041762
Acid Blue 83, C.I. 42660, Coomassie™ Brilliant Blue R-250
sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-060-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only