missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Blue R

CAS: 6104-59-2 | C45H44N3NaO7S2 | 825.97 g/mol

Supplier:  Thermo Scientific Chemicals J6429718

Catalog No. AAJ6429718


Only null left
Explore available promotions

Description

Description

Brilliant Blue R is used for detection of protein bands following electrophoresis. It is a sensitive stain for protein detection in polyacrylamide gels, typically giving blue bands on a clear background with a sensitivity of 50-100 ng/band. It may be combined with other stains, such as silver stain, to distinguish different types of proteins. It displays metachromasia by appearing pink-red, rather than blue, when binding certain proteins, such as collagen and histone H1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Blue R is used for detection of protein bands following electrophoresis. It is a sensitive stain for protein detection in polyacrylamide gels, typically giving blue bands on a clear background with a sensitivity of 50-100
Specifications

Specifications

Brilliant Blue R
C45H44N3NaO7S2
5718025
Soluble in water.
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
825.97
825.97
50 g
6104-59-2
MFCD00041762
Coomassie Brilliant Blue R 250; C.I. 42660
HAEGGLCXLFWTBE-UHFFFAOYSA-M
sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
61365
Blue to Red
Crystalline Powder

Chemical Identifiers

6104-59-2
825.97
HAEGGLCXLFWTBE-UHFFFAOYSA-M
61365
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
C45H44N3NaO7S2
MFCD00041762
Coomassie Brilliant Blue R 250; C.I. 42660
sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 228-060-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only