missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Blue G, pure

CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol

$50.14 - $272.27

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
Molecular Weight (g/mol) 854.025
MDL Number MFCD00078482
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Synonym Acid Blue 90, Brilliant Blue G 250, C.I. 42655
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC191480050
SDS View Documents
Thermo Scientific Chemicals
191480050
5 g Glass bottle
Each for $50.14
Only null left
 
AC191480250
SDS View Documents
Thermo Scientific Chemicals
191480250
25 g Glass bottle
Each for $106.17
Only null left
 
AC191481000
SDS View Documents
Thermo Scientific Chemicals
191481000
100 g Glass bottle
Each for $272.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6104-58-1
854.025
RWVGQQGBQSJDQV-UHFFFAOYSA-M
6328534
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
C47H48N3NaO7S2
MFCD00078482
Acid Blue 90, Brilliant Blue G 250, C.I. 42655
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate

Specifications

425 (at lambda min.)
6104-58-1
60% min.
MFCD00078482
Acid Blue 90, Brilliant Blue G 250, C.I. 42655
RWVGQQGBQSJDQV-UHFFFAOYSA-M
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]
854.025
608 to 618nm (in 50:50 IMS:H2O)
854.02
Glass bottle
Crystalline Powder
>100.0°C
0.95 to 1.15
C47H48N3NaO7S2
14, II, 450
Solubility in water: 40g/L (20°C). Other solubilities: soluble in methanol, ethanol
Authentic
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
6328534
10% max. (105°C)
Pure
5 g
Brilliant blue G
Videos
SDS
Documents

Documents

RUO – Research Use Only