missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Blue G, pure

Catalog No. AC191480050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191480050 5 g Glass bottle
AC191480250 25 g Glass bottle
AC191481000 100 g Glass bottle
AC191485000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC191480050 Supplier Thermo Scientific Chemicals Supplier No. 191480050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
Molecular Weight (g/mol) 854.025
MDL Number MFCD00078482
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Synonym Acid Blue 90, Brilliant Blue G 250, C.I. 42655
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

Absorbance 425 (at lambda min.)
Melting Point >100.0°C
Absorption Ratio 0.95 to 1.15
Dye Content 60% min.
Beilstein 14, II, 450
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: soluble in methanol, ethanol
Infrared Spectrum Authentic
Lambda Max 608 to 618nm (in 50:50 IMS:H2O)
Formula Weight 854.02
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Glass bottle
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Brilliant blue G
Show More Show Less

RUO – Research Use Only