missing translation for 'onlineSavingsMsg'
Learn More

Brij™ C20

CAS: 9004-95-9 | C56H114O21 | 1123.51 g/mol

$70.02 - $688.76

Chemical Identifiers

CAS 9004-95-9
Molecular Formula C56H114O21
Molecular Weight (g/mol) 1123.51
MDL Number MFCD00080892
InChI Key NLMKTBGFQGKQEV-UHFFFAOYSA-N
Synonym Polyoxyethylene(20) cetyl ether
PubChem CID 2724259
SMILES CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC344290050
View Documents Promotion Details
N/A 5 g
Each for $70.02
Only null left
 
AC344295000
View Documents Promotion Details
N/A 500 g
Each for $166.93
Only null left
 
AC344290025
View Documents Promotion Details
N/A 2.5 kg
Each for $688.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9004-95-9
1123.51
NLMKTBGFQGKQEV-UHFFFAOYSA-N
2724259
C56H114O21
MFCD00080892
Polyoxyethylene(20) cetyl ether
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

White
1mg KOH/g max.
35.4°C to 39.6°C
45 to 60mg KOH/g
Glass bottle
C16H33(OCH2CH2)20OH
MFCD00080892
Polyoxyethylene(20) cetyl ether
NLMKTBGFQGKQEV-UHFFFAOYSA-N
2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-hexadecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
2724259
Brij™ C20
9004-95-9
Pellets
>149°C
Authentic
C56H114O21
1% max.
10, 7449
Solubility in water: soluble. Other solubilities: soluble in lower alcohols, insoluble in propylene glycol, coal-tar hydrocarbo, n, cottonseed and mineral oil
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
1123.51
1123.5
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TR1581470

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only