missing translation for 'onlineSavingsMsg'
Learn More

Brij™ 98

Catalog No. p-71992
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347181000 100 g Glass bottle
AC347185000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC347181000 Supplier Thermo Scientific Chemicals Supplier No. 347181000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9004-98-2 | C58H116O21 | 1149.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-98-2
Molecular Formula C58H116O21
Molecular Weight (g/mol) 1149.55
MDL Number MFCD00074369
InChI Key TVSFHLHLHUYZNN-KTKRTIGZSA-N
Synonym Polyoxyethylene(20) oleyl ether
PubChem CID 5364713
IUPAC Name (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White to Cream
Acid Value 1mg KOH/g max.
Physical Form Soft Waxy Solid
Boiling Point >100.0°C
Melting Point 30.0°C
Density 1.0700g/mL
Quantity 100 g
Flash Point >150°C
Hydroxyl Value 50 to 65mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.07
Linear Formula C18H35(OCH2CH2)nOH
Water 3% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, toluene, propylene glycol, insoluble in cottonseed oil, mineral oil
Formula Weight 1149.56
Chemical Name or Material Brij™ 98
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber RK3150000
TSCA TSCA

RUO – Research Use Only