Learn More
Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35
CAS: 9002-92-0 | C58H118O24
Supplier: Thermo Scientific Chemicals 329580010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| Colorless | |
| Jelly Substance at 25°C | |
| Glass bottle | |
| C58H118O24 | |
| MFCD00080891 | |
| 15, 7675 | |
| 1199.56 | |
| Brij™ 35 |
| 9002-92-0 , 7732-18-5 | |
| 1.0300g/mL | |
| 1.03 | |
| C12H25(OCH2CH2)23OH | |
| 06,70 | |
| Solubility in water: soluble. | |
| 29 to 31% |
Specifications
| Colorless | |
| Jelly Substance at 25°C | |
| 1 L | |
| 1.03 | |
| C12H25(OCH2CH2)23OH | |
| 06,70 | |
| Solubility in water: soluble. | |
| 29 to 31% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only