BrightStar™ is a white, light-reflecting, vinyl Sealing film that can be applied to either the top or bottom of the plate. White, light-reflecting vinyl film that can be applied to either the top or bottom of the plate. The light reflective properties on clear-bottom plates maximize the sensitivity of bioluminescence and chemiluminescence assays in bottom-reading luminometers. Maximizes intensity and signal-to-noise ratios in luminescence assays. May be used as a backing matrix for microscopy of punchout samples for filter plates (e.g. ELISPOT assays). Features a single split liner for easy application .