missing translation for 'onlineSavingsMsg'
Learn More

BrettPhos Palladacycle 3rd Generation, 98%

CAS: 1470372-59-8

$238.32 - $3398.22

Chemical Identifiers

CAS 1470372-59-8
MDL Number MFCD22572666
Synonym [(BrettPhos)-2-(2’-amino-1,1’-biphenyl)]palladium (II) methanesulfonate; BrettPhos Pd G3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA47250MD
View Documents
Thermo Scientific Chemicals
047250MD
250 mg
Each for $238.32
Only null left
 
AA4725003
View Documents
Thermo Scientific Chemicals
04725003
1 g
Each for $902.07
Only null left
 
AA4725006
View Documents
Thermo Scientific Chemicals
04725006
5 g
Each for $3,398.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1470372-59-8
MFCD22572666
[(BrettPhos)-2-(2’-amino-1,1’-biphenyl)]palladium (II) methanesulfonate; BrettPhos Pd G3

Specifications

1470372-59-8
MFCD22572666
[(BrettPhos)-2-(2’-amino-1,1’-biphenyl)]palladium (II) methanesulfonate; BrettPhos Pd G3
98%
BrettPhos Palladacycle 3rd Generation
Yellow
250 mg
906.5
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only