missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Brefeldin A, natural
CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol
Supplier: Thermo Scientific Chemicals 297140250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Brefeldin A | |
| 201.0°C to 205.0°C | |
| Authentic | |
| C16H24O4 | |
| 25 mg | |
| + 93.00 | |
| Solubility in water: .. Other solubilities: 10 mg/mL methanol | |
| C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1 | |
| 280.36 | |
| 280.35 | |
| Crystalline Powder |
| 20350-15-6 | |
| White | |
| Glass bottle | |
| MFCD00083258,MFCD12913297 | |
| 15, 1374 | |
| brefeldin a, cyanein, decumbin, bfa | |
| KQNZDYYTLMIZCT-PNFJWZTBSA-N | |
| (1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one | |
| 133640201 | |
| ≥98.0% |
Chemical Identifiers
| 20350-15-6 | |
| 280.36 | |
| KQNZDYYTLMIZCT-PNFJWZTBSA-N | |
| 133640201 | |
| C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1 |
| C16H24O4 | |
| MFCD00083258,MFCD12913297 | |
| brefeldin a, cyanein, decumbin, bfa | |
| (1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one |
Safety and Handling
RTECSNumber : GY8410000
RUO – Research Use Only