missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brefeldin A, natural

Catalog No. AC297140050
Change view
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297140050 5 mg Glass bottle
AC297140250 25 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297140050 Supplier Thermo Scientific Chemicals Supplier No. 297140050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4
Molecular Weight (g/mol) 280.36
MDL Number MFCD00083258,MFCD12913297
InChI Key KQNZDYYTLMIZCT-PNFJWZTBSA-N
Synonym brefeldin a, cyanein, decumbin, bfa
PubChem CID 133640201
IUPAC Name (1S,7S,10E,12R,13R,15S)-12,15-dihydroxy-7-methyl-8-oxabicyclo[11.3.0]hexadeca-2,10-dien-9-one
SMILES C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

Specifications

Melting Point 201.0°C to 205.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 mg
Merck Index 15, 1374
Specific Rotation + 93.00
Solubility Information Solubility in water: .. Other solubilities: 10 mg/mL methanol
Formula Weight 280.35
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material Brefeldin A
Show More Show Less
RTECSNumber GY8410000

RUO – Research Use Only