accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Back to School
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Cell Culture Media, Supplements, and Reagents
Cell Culture Reagents, Antibiotics and Supplements
Antibiotics
ICN15902705
Structure Search
Print
Brefeldin A, MP Biomedicals
Catalog No.
ICN15902705
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
ICN15902705
5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
ICN15902705
Supplier
MP Biomedicals Inc
Supplier No.
0215902705
Sign In
or
Register
to check your price and availability.
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
20350-15-6
Molecular Formula
C16H24O4
Molecular Weight (g/mol)
280.36
MDL Number
MFCD00083258,MFCD12913297
InChI Key
KQNZDYYTLMIZCT-PNFJWZTBSA-N
PubChem CID
133640201
IUPAC Name
(1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
SMILES
C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1
Specifications
Quantity
5 mg
Product Suggestions
Documents