missing translation for 'onlineSavingsMsg'
Learn More

Brefeldin A, 98%

Catalog No. p-7042386
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62340MA 10 mg
AAJ62340MB 25 mg
AAJ62340MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62340MA Supplier Thermo Scientific Chemicals Supplier No. J62340MA

CAS: 20350-15-6 | C16H24O4 | 280.36 g/mol

Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brefeldin A mediated apoptosis has been observed in human tumor cellsIt is an antibiotic used in cell culture and to inhibit protein secretion in mammalian and other eukaryotic cells by interfering with the function of the Golgi apparatus.

Solubility
Soluble in dimethylsulfoxide, dichloromethane and ethanol. Slightly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 20350-15-6
Molecular Formula C16H24O4
Molecular Weight (g/mol) 280.36
MDL Number MFCD00083258,MFCD12913297
InChI Key KQNZDYYTLMIZCT-PNFJWZTBSA-N
Synonym brefeldin a, cyanein, decumbin, bfa
PubChem CID 133640201
SMILES C[C@H]1CCC\C=C\[C@@H]2C[C@H](O)C[C@H]2[C@@H](O)\C=C\C(=O)O1

Specifications

Melting Point 201°C to 205°C
Color White
Quantity 10 mg
UN Number UN2811
Beilstein 25191
Merck Index 14,1369
Solubility Information Soluble in dimethylsulfoxide,dichloromethane and ethanol. Slightly soluble in water.
IUPAC Name (1S,6S,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-1H,4H,6H,7H,8H,9H,11aH,12H,13H,14H,14aH-cyclopenta[f]oxacyclotridecan-4-one
Formula Weight 280.36
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brefeldin A
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber GY8410000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only