missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brassinazole 93.0+%, TCI America™
$46.60 - $270.29
Chemical Identifiers
| CAS | 224047-41-0 |
|---|---|
| Molecular Formula | C18H18ClN3O |
| Molecular Weight (g/mol) | 327.81 |
| MDL Number | MFCD08276319 |
| InChI Key | YULDTPKHZNKFEY-UHFFFAOYNA-N |
| Synonym | 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol |
| PubChem CID | 15477807 |
| ChEBI | CHEBI:73177 |
| IUPAC Name | 4-(4-chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol |
| SMILES | CC(O)(C(CC1=CC=C(Cl)C=C1)N1C=NC=N1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B282910MG
|
TCI America
B282910MG |
10 mg |
Each for $46.60
|
|
|||||
|
B2829100MG
|
TCI America
B2829100MG |
100 mg |
Each for $270.29
|
|
|||||
Chemical Identifiers
| 224047-41-0 | |
| 327.81 | |
| YULDTPKHZNKFEY-UHFFFAOYNA-N | |
| 15477807 | |
| 4-(4-chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol |
| C18H18ClN3O | |
| MFCD08276319 | |
| 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol | |
| CHEBI:73177 | |
| CC(O)(C(CC1=CC=C(Cl)C=C1)N1C=NC=N1)C1=CC=CC=C1 |
Specifications
| 224047-41-0 | |
| C18H18ClN3O | |
| 10 mg | |
| YULDTPKHZNKFEY-UHFFFAOYNA-N | |
| 4-(4-chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol | |
| 15477807 | |
| 327.81 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD08276319 | |
| 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol | |
| CC(O)(C(CC1=CC=C(Cl)C=C1)N1C=NC=N1)C1=CC=CC=C1 | |
| 327.81 | |
| CHEBI:73177 | |
| ≥93.0% (HPLC) | |
| Brassinazole |
Safety and Handling
TSCA : No