missing translation for 'onlineSavingsMsg'
Learn More

Brassinazole 93.0+%, TCI America™

Catalog No. B282910MG
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B282910MG 10 mg
B2829100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B282910MG Supplier TCI America Supplier No. B282910MG
Only null left

Chemical Identifiers

CAS 224047-41-0
Molecular Formula C18H18ClN3O
Molecular Weight (g/mol) 327.81
MDL Number MFCD08276319
InChI Key YULDTPKHZNKFEY-UHFFFAOYNA-N
Synonym 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol
PubChem CID 15477807
ChEBI CHEBI:73177
IUPAC Name 4-(4-chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(O)(C(CC1=CC=C(Cl)C=C1)N1C=NC=N1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 327.81
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Brassinazole