missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brass foil, alloy 260, 2.0mm (0.08in) thick
$171.23 - $171.23
Specifications
| Chemical Name or Material | Brass foil, Alloy 260 |
|---|---|
| Name Note | 2.0mm (0.08 in.) thick |
| Odor | Odorless |
| Form | Foil |
| MDL Number | MFCD00171255 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Brass foil, Alloy 260 | |
| Odorless | |
| MFCD00171255 |
| 2.0mm (0.08 in.) thick | |
| Foil |
RUO – Research Use Only