Learn More
Eisco™ Brass Cork Borer Set - German Pattern
Useful for cutting a hole in cork or rubber stoppers to insert glass tubing.
Supplier:  Eisco™ CH0322B
Catalog No. S27339
Only null left
Description
Description
  • Set of 12 cork borers with sizes ranging from 4 to 18 mm
  • Brass construction with a nickel plated finish
  • Cleaning rod included
  • German Pattern
Specifications
Specifications
Cork Borers
4 to 18 mm
German Pattern
12 Borers
Cork Borer
Brass
Cutting a hole in cork or rubber stoppers to insert glass tubing
Product Suggestions
Product Suggestions