Learn More
Branson Ultrasonics™ Sonifier™ Replacement Flat Tip
Description
Tip to use as a 1/2 in. probe, without tip probe used to attach tapered microtips.
Specifications
Specifications
| For Use With (Equipment) | Sonifier™ cell disruptor |
| Tip Style | Flat |
| Product Type | Flat Microtip |