Learn More
Branson Ultrasonics™ Ultrasonic Cleaning Bath Accessory, Covers
For use with Bransonic cleaning baths
Supplier:  Branson Ultrasonics™ 100032519
 View more versions of this product
Catalog No. 15345128
Only null left
Description
Description
For use with Bransonic cleaning baths
  • Stainless steel lid
Specifications
Chemical Identifiers
Lid
Helps position beakers securely in bath
Specifications
Lid
Ultrasonic Bath, Model 3800
Helps position beakers securely in bath
Plastic
Product Suggestions
Product Suggestions