Learn More
BRANDTECH™ VITLAB™ Measuring Scoops
In assorted sizes
Supplier:  BRANDTECH™ VITLAB™ 40793
 View more versions of this product
Catalog No. 14-379-290
Only null left
Explore available promotions
Description
Description
  • Choose polypropylene or polyethylene
  • Metric volume (mL) marked on handle
Specifications
Specifications
Not Autoclavable
1250 mL
For measuring samples
HDPE
42.26 oz.
Measuring Scoop
101.6 in.
40 cm
Product Suggestions
Product Suggestions