Learn More
BRANDTECH™ VACUUBRAND™ Rotary Vane Vacuum Pumps: Inlet Separators
Inlet separators for VACUUBRAND™ Rotary Vane Vacuum Pumps.
Supplier: BRANDTECH™ VACUUBRAND™ 20698007
Description
- For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
- Inlet Separators 'AK' help keep condensates and particulates from entering the vacuum pump, preserving performance.
Note: For RE/RZ 2.5: For rotary vane pumps RE 2.5 (article no. 2079715x) and RZ 2.5 (article no. 2079812x) - please order Inlet Separator 20698006. For rotary vane pumps RE 2.5 (article no. 2069715x) and RZ 2.5 (article no. 2069812x) - please order Inlet Separator 20698000.
Chemical Identifiers
| To keep condensates and particulates from entering the vacuum pump | |
| Inlet Catchpot/Separator |
Specifications
| To keep condensates and particulates from entering the vacuum pump | |
| RE8, RE9, RE16, RZ8, RZ9, RZ16 | |
| Inlet Catchpot/Separator |