Learn More
BRANDTECH™ VACUUBRAND™ Rotary Vane Vacuum Pumps: Inlet Separators
Inlet separators for VACUUBRAND™ Rotary Vane Vacuum Pumps.
Supplier:  BRANDTECH™ VACUUBRAND™ 20698000
 View more versions of this product
Catalog No. 13889109
Only null left
Description
Description
  • For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
  • Inlet Separators 'AK' help keep condensates and particulates from entering the vacuum pump, preserving performance.
Note: For RE/RZ 2.5: For rotary vane pumps RE 2.5 (article no. 2079715x) and RZ 2.5 (article no. 2079812x) - please order Inlet Separator 20698006. For rotary vane pumps RE 2.5 (article no. 2069715x) and RZ 2.5 (article no. 2069812x) - please order Inlet Separator 20698000.
Specifications
Chemical Identifiers
To keep condensates and particulates from entering the vacuum pump
Inlet Catchpot/Separator
Specifications
To keep condensates and particulates from entering the vacuum pump
RE2, RZ2, RE 2.5, RZ 2.5
20698000
Inlet Catchpot/Separator
Product Suggestions
Product Suggestions